Learn More
Alkali Scientific 0.6mL Microcentrifuge Tubes, Lock Cap, Clear, Conical, DNase, RNase & Pyrogen Free, 10000/Cs
SDP
Supplier:  Alkali Scientific CN3009
Encompass_Preferred
Our centrifuge tubes could be commonly used to collect cells and separate biomacromolecules by centrifugation in biochemistry and other biological fields. They can also be used to store photosensitive samples and special chemiluminescence reagents. USP VI grade polypropylene material, free from heavy metal. RCF: 30,000 xg. Special safe lock cap design provides secure lid sealing. Clear graduations and frosted marking area. Easy to open and close. Autoclavable. RNase free, endotoxin level less than 0.1EU.
Catalog No. 50-006-62631