Learn More
Azenta US Inc 0.9ML DUAL-CODED TUBE, 960/CS
Supplier:  Azenta US Inc 68100100
Encompass
0.9ml Dual-coded Tube, 96-format, External Thread, 2D Codeand Human Readable Number on base, uncapped, bulk, 960 tubes per case
Catalog No. NC2199110