Learn More
Refrigiwear Inc. 0025SG Regular Clear OSA Anti-Fog Safety Glasses
SDP
Supplier:  Refrigiwear Inc. 0025SG00000
Encompass_Preferred
Premium Safety Glasses are the best in comfort and protection. They meeting multiple standards, have anti-fog protection, and ANSI-approved hard coated polycarbonate lenses. The removeable EVA foam dust filter around the eyes keeps dust out while letting air in. No metal components makes these great for metal-free environments. Meets ANSI Z87.1 +,EN166K, EN166N, EN166UV requirements Clear vision with Brass Knuckle N-FOG anti-fog protection that exceeds European EN 166/168 standards Removeable, soft EVA foam dust filter with built-in air channels fills the gap between the glasses and the face, keeping dust out comfortably ANSI-approved hardcoated polycarbonate lenses Molded nosepiece for all-day comfort No metal components
Catalog No. NC3867020