Learn More
ACCUTEC BLADES INC (3P) .009" Carbon Steel 2-Facet Degreased Blades, Steel Back, Bulk, 100 Blades per Cartridge, 40 Cartridges per Case
Supplier:  ACCUTEC BLADES INC (3P) AGBL7051BULK
Encompass_Preferred
AccuForge Single Edge blades feature .009 high carbon steel for durability, and standard 2-Facet edges. Blades are degreased to remove residual oil. Each cartridge contains 100 blades. Bulk pack of 40 cartridges. Fits Gem Runner™ Dispenser AGTL-7000-0000. Made in the USA
Catalog No. NC3721084