Learn More
ACCUTEC BLADES INC (3P) .009" Stainless Steel 3-Facet MicroCoat Degreased Blades, Aluminum Back, Unwrapped, 115 Blades per Clamshell, 50 Clamshells per Case
Supplier:  ACCUTEC BLADES INC (3P) AGBL70220000
Encompass_Preferred
AccuForge Single Edge blades feature .009 stainless steel and 3-facet edges for sharpness. Blades are degreased to remove residual oil. Our proprietary MicroCoat Enrichment Technology reduces cut force and helps the blade maintain a sharp effective edge over multiple uses. Each clamshell contains 115 unwrapped blades. Made in the USA.
Catalog No. NC3721101