Learn More
MSC Value Collection Plug Gage Set, 0.011-0.06 in, 50 Pc
Supplier: MSC 05046594
This Class ZZ Plug Gage Set offers precise measurement for various applications. It includes 50 pieces, covering a diameter range of 0.011 to 0.06 in. with a tolerance of 0.000200 in..
- Precise measurement capability
- Contains 50 individual plug gages
- Designed for Class ZZ tolerance type
- Ideal for checking small hole diameters