Learn More
Alkali Scientific 0.75 Liter Lab Armor Metal Beads for Bead Bath
SDP
Supplier:  Alkali Scientific WB523
Encompass
0.75 Liter Lab Armor Metal Beads have a metal composition, are moisture and gas impermeable. They have high thermal conductivity, and a smooth, rounded surface. The working temperature range is -80°C to 180°C. They improve energy efficiency of standard water baths by over 50%.
Catalog No. 50-233-5990