Learn More
Accuflow Systems C-Flex Tubing, Saint Gobain, 082, TPE, thermoplastic elastomer, weldable, 50ft
Supplier:  Accuflow Systems 08210002
Encompass_Preferred
C-Flex Tubing, Saint Gobain, 082, TPE, thermoplastic elastomer, weldable, 50ft
Catalog No. 50-252-490