Learn More
World Precision Instrument 1.0 mm Glass Pipette Holder for NANOFIL Syringe
SDP
Supplier:  World Precision Instrument NFINHLDG10
Encompass
A kit to use with the NANOFIL syringe and 1mm OD glass for your microinjection application. 
Catalog No. 50-252-1060