Learn More
Glen Mills Inc Glass Beads 1.00 - 1.30, 1lb
SDP
Supplier:  Glen Mills Inc 7200001000
Encompass
These soda-lime lead-free glass beads are extremely round, have a narrow size spread, and their durability makes them ideal for thinner suspensions such as agricultural chemicals, tape coatings, aqueous suspensions, biologicals, and many other milling operations. *This item is for 1 pound -Beads purchased at less than full bag (44 pounds), have a $25.00 handling fee.
Catalog No. NC2086750