Learn More
Medchemexpress LLC 1,1′,1′′,1′′′-[1,4-Piperazinediylbis(2,1-ethanediylnitrilo)]tetrakis[2-dodecanol] | 1265904-26-4 | 98.0% | 909.54 | 1 ML

Supplier: Medchemexpress LLC HY11277310MM/1ML
1,1′,1′′,1′′′-[1,4-Piperazinediylbis(2,1-ethanediylnitrilo)]tetrakis[2-dodecanol] is a lipid/lipidoid utilized in the preparation of lipid-based or lipidoid nanoparticles. It is intended for research use only and is not sold to patients.
- Appearance: Solid
- Color: Light yellow to orange
- Shipping: Room temperature in continental US; may vary elsewhere
- Storage (powder): -20°C for 3 years; 4°C for 2 years
- Storage (in solvent): -80°C for 6 months; -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.