Learn More
Medchemexpress LLC 1,1′,1′′,1′′′-[1,4-Piperazinediylbis(2,1-ethanediylnitrilo)]tetrakis[2-dodecanol] | 1265904-26-4 | 98.0% | 909.54 | 10 MG

Supplier: Medchemexpress LLC HY11277310MG
1,1′,1′′,1′′′-[1,4-Piperazinediylbis(2,1-ethanediylnitrilo)]tetrakis[2-dodecanol] is a lipid/lipidoid for the preparation of lipid-based or lipidoid nanoparticles. This product is for research use only and not sold to patients.
- Used in preparation of lipid-based or lipidoid nanoparticles.
- Appearance as a solid, light yellow to orange.
- Recommended storage for powder is -20°C for 3 years, 4°C for 2 years.
- Recommended storage for in solvent is -80°C for 6 months, -20°C for 1 month.
- Soluble in DMSO (25 mg/mL), requiring sonication.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.