Learn More
Medchemexpress LLC 1-(1,1-dimethylethyl) tridecanedioate | 2703752-86-5 | 300.43 g/mol | C17H32O4 | 1g
SDP
Supplier:  Medchemexpress LLC HYW8814391G
Encompass_Preferred
1-(1 1-Dimethylethyl) tridecanedioate is a PROTAC Linker and can be used for PROTAC synthesis such as FF2039 (HY-172159)[1]
Catalog No. 50-002-69589