Learn More
Matrix Scientific 1,1,1-Trifluoro-2,3-epoxypropane, 359-41-1, MFCD00041506, 5g
Supplier: Matrix Scientific 0035635G
Molecular Formula C3H3F3O, Purity 98%, Molecular Weight 112.05, Boiling Point 39
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.