Learn More
Medchemexpress LLC 1,1,1-Trifluoroethyl-PEG2-azide | 1835759-68-6 | 97.0% | C6H10F3N3O2 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY140845
Encompass_Preferred
1,1,1-Trifluoroethyl-PEG2-azide is a PEG-based PROTAC linker used in the synthesis of PROTACs. It acts as a click chemistry reagent, capable of participating in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions.
  • Contains an azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with alkyne groups
  • Can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups
Catalog No. 50-003-52729