Learn More
Medchemexpress LLC 1,1,1-Trifluoroethyl-PEG2-propargyl | 1835759-73-3 | ≥97.0% | C9H13F3O3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140886100MG
Encompass_Preferred
1,1,1-Trifluoroethyl-PEG2-propargyl is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, possessing an Alkyne group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing Azide groups. This design enables PROTACs to leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent with alkyne group
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Enables selective degradation of target proteins via ubiquitin-proteasome system
Catalog No. 50-003-55112