Learn More
Chemglass Life Sciences Chemglass 1-1/2 in Sanitary Non-Valved MPC Coupling Insert, Polysulfone, 25 per pack
SDP
Supplier:  Chemglass Life Sciences CLS91419024
Encompass
Chemglass sanitary non-valved MPC coupling insert designed for quick-connect sanitary fluid handling. Constructed primarily from USP Class VI polysulfone with optional locking sleeves and multiple O-ring material options, this insert fits common tubing IDs and is supplied in packs of 25 for assembly or replacement.
  • Fits tubing inner diameters 1/8 in, 1/4 in, and 3/8 in.
  • Main component made of polysulfone, USP Class VI, ADCF.
  • Locking sleeves made of polysulfone for secure connections.
  • Thumb latches available in polycarbonate or PVDF.
  • O-rings available in silicone (platinum-cured) and Buna-N.
  • Pack quantity 25 per pack.
  • Coupling halves can be rotated when connected to reduce tube kinks.
  • Suitable for critical fluid handling applications.
Catalog No. NC2547879