Learn More
Chemglass Life Sciences 1-1/2" Sanitary Genderless AseptiQuik G Connector Body
SDP
Supplier:  Chemglass Life Sciences CLS91307024
Encompass
Material: Polycarbonate, USP Class VI, ADCF. Material Finish: Molded White. Pressure Range: Up to 60 psi, 4. 1 bar for 30 days; Up to 75 psi, 5. 1 bar for 48 hours. Color: White with blue pull tab. Mounting Option: Free Floating. Supplied Non-Sterile: Gamma Radiation Compatible. Temperature Range: 34 - 104 ° F (1 - 40 ° C). Seal Material: Silcone, Platinum-cured, USP Class VI, ADCF. Valve Option: Non-Valved.
Catalog No. 50-009-80606