Learn More
Medchemexpress LLC 1-(1,3-benzothiazol-2-yl)-1-(2,4-dimethylphenyl)ethanol | 1253901-26-6 | MFCD20527324 | >98.0% | 283.39 g/mol | C17H17NOS | 5 MG

Supplier: Medchemexpress LLC HY1178515MG
1-(1,3-benzothiazol-2-yl)-1-(2,4-dimethylphenyl)ethanol is a calcium-sensing receptor (CaSR) agonist and positive allosteric modulator supplied as a 5 mg research reagent for in vitro and in vivo studies of CaSR signaling and related disease models. The compound is provided as a solid with documented solubility and storage recommendations.
- Purity ≥98% (HPLC)
- Molecular weight 283.39 g/mol
- Chemical formula C17H17NOS
- Soluble in DMSO (100 mg/mL); in vivo formulations achieve ≥2.5 mg/mL as clear solutions
- Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month)
- Target: calcium-sensing receptor (CaSR); pathway: GPCR/G protein
- For research use only; not for human or diagnostic use
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.