Learn More
Medchemexpress LLC 1,1':4',1''-terphenyl | 92-94-4 | MFCD00003061 | 99.8% | 230.31 g/mol | C18H14 | 100 G

Supplier: Medchemexpress LLC HYW013973100G
p-Terphenyl (CAS 92-94-4) is an aromatic hydrocarbon used as a research chemical, synthetic intermediate, and analytical reference. It has formula C18H14 and a molecular weight of 230.31 g/mol. Derivatives have reported anticancer and anti-inflammatory activity. The material is supplied as a powder with supporting analytical data and storage recommendations.
- Molecular formula C18H14 and molecular weight 230.31 g/mol.
- High purity, about 99.8% by HPLC.
- Supplied as a solid powder suitable for synthesis and analysis.
- Includes analytical documents such as COA, SDS, 1H NMR, and RP-HPLC.
- Recommended storage: powder at -20°C for long-term or 4°C for shorter-term storage.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.