Learn More
Medchemexpress LLC [1,1'-Biphenyl]-2,2'-dicarboxylic acid, 4,4'-dimethyl- | 2941-79-9 | ≥98.0% | C16H14O4 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY78572
Encompass_Preferred
Suvorexant impurity 1 is a chemical compound with a molecular formula of C16H14O4 and a CAS number of 2941-79-9. It is provided as an off-white to brown solid with a purity of ≥98.0%. This product is tested and complies with given specifications.
  • Off-white to brown solid appearance
  • Molecular weight: 270.28
  • Consistent with structure by 1H NMR spectrum
  • Stable as powder at -20°C for 3 years or 4°C for 2 years
  • Stable in solvent at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-003-59253