Learn More
Medchemexpress LLC [1,1′-Biphenyl]-2-amine, 2′-(dicyclohexylphosphino)-N,N-dimethyl- | 213697-53-1 | 393.54 | 10 G

Supplier: Medchemexpress LLC HYW00292510G
DavePhos is a biochemical reagent designed for life science research. It can be utilized as a biological material or organic compound in various research applications. This product is intended for research use only and is not sold for medical applications or to patients.
- Off-white to light yellow solid
- Storage at 4°C, protected from light and under nitrogen
- In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light and under nitrogen
- Soluble in DMF at 6.67 mg/mL (16.95 mM)
- Purity of 98.0% or greater
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.