Learn More
Medchemexpress LLC [1,1'-Biphenyl]-4-propanoic acid, α-amino-, (S)- | 155760-02-4 | 99.9% | C15H15NO2 | 5 G
SDP
Supplier:  Medchemexpress LLC HY791065G
Encompass_Preferred
L-Biphenylalanine is a phenylalanine derivative commercially used as an ergogenic supplement. It is recognized for its beneficial effects on physical, mental, and physiological activities, including influencing anabolic hormone secretion, supplying fuel during exercise, enhancing mental performance, and preventing exercise-induced muscle damage.
  • Influences anabolic hormone secretion.
  • Supplies fuel during exercise.
  • Enhances mental performance during stress-related tasks.
  • Prevents exercise-induced muscle damage.
Catalog No. 50-004-35035