Learn More
Medchemexpress LLC (+)-1-(1-Naphthyl)ethylamine | 3886-70-2 | 99.99% | C12H13N | 100 G
SDP
Supplier:  Medchemexpress LLC HY75070100G
Encompass_Preferred
(+)-1-(1-Naphthyl)ethylamine ((+)-1-(1-NEA)) is a chiral modifier utilized to introduce enantioselectivity in catalytic hydrogenation reactions. It can undergo H-D exchange with D2 in solution to form N-D bonds. This indicates that 1-NEA can complete Pt surface adsorption and protonation through amine N atoms, suggesting that NEA molecules have the potential to impart enantioselectivity to Pt hydrogenation catalysts.
  • Chiral modifier
  • Introduces enantioselectivity in catalytic hydrogenation reactions
  • Undergoes H-D exchange with D2 in solution to form N-D bonds
  • Suggests potential to impart enantioselectivity to Pt hydrogenation catalysts
Catalog No. 50-004-34368