Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-1(Z)-Hexadecenyl-2-Palmitoyl-sn-glycero-3-PC | 126901-45-9 | 97.1% | 718.04 | 1 MG
SDP

Supplier:  Medchemexpress LLC HY1659971MG

Encompass_Preferred

1-1(Z)-Hexadecenyl-2-Palmitoyl-sn-glycero-3-PC is an ester product provided for research use only. This compound is available in solid form, appearing white to off-white, and has a molecular weight of 718.04. It is identified by CAS number 126901-45-9 and boasts a purity of 97.1%.

  • Ester product
  • High purity (97.1%)
  • Available in various quantities (e.g., 1 mg)
  • Ships at room temperature within the continental US
  • Long shelf life when stored properly (Powder: -20°C for 3 years; In solvent: -80°C for 6 months, -20°C for 1 month)

Catalog No. 50-003-94495


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.