Learn More
Medchemexpress LLC 1,10-decanediol | 112-47-0 | MFCD00004749 | 99.9% | 174.28 g/mol | C10H22O2 | 500 G

Supplier: Medchemexpress LLC HYW016773500G
1,10-Decanediol is a linear diol (decane-1,10-diol) with two terminal primary hydroxyl groups, used as a building block in polymer synthesis, formulation chemistry, and materials research. It is commonly used as a bifunctional spacer in condensation reactions and crosslinking for producing polyesters and related materials.
- Bifunctional primary diol with terminal hydroxyl groups.
- Chemical formula C10H22O2; molecular weight 174.28 g/mol.
- CAS number 112-47-0 for unambiguous identification.
- High purity (≈99.9%) as supplied.
- Stable at room temperature; storage stability listed as 3 years.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.