Learn More
Research Products International Corp 1, 10-Phenanthroline, 50 Grams
SDP
Supplier:  Research Products International Corp P3020050.0
Encompass_Preferred
Forms a complex with ferrous ions. It can be used as an indicator in oxidation-reduction systems in fitrating ferrous salts. Material passes test for redox indicator and passes test for iron determination.
Catalog No. 50-488-910