Learn More
Medchemexpress LLC 1,11-Bis(methanesulfonyloxy)-3,6,9-trioxandecane | 55400-73-2 | 95.0% | 350.41 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY42774
Encompass_Preferred
Ms-PEG4-MS is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Used in research only, not sold to patients
Catalog No. 50-004-03278