Learn More
Medchemexpress LLC 1-[11-(dodecylamino)-10-hydroxyundecyl]-3,7-dimethylpurine-2,6-dione | 166981-13-1 | 99.4% | 533.79 g/mol | C30H55N5O3 | 10 MG

Supplier: Medchemexpress LLC HY11898210MG
CT-2584 is a chemotherapeutic research compound that reduces NKEF-B expression and kills tumor cells by inducing mitochondrial reactive oxygen species (ROS). It is used in preclinical cancer studies to probe phospholipid signaling and cytotoxic mechanisms.
- Chemotherapeutic compound that induces mitochondrial ROS.
- Reduces NKEF-B expression in tumor cell lines.
- High chemical purity (99.4%).
- Available as a solid, 10 mg package for laboratory use.
- Soluble in DMSO and ethanol with recommended handling conditions.
- Storage stability specified for frozen and refrigerated conditions.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.