Learn More
Chemglass Life Sciences 1/16" Dia X 12" Long PTFE Coated Thermocouple Probe Type "J"
SDP
Supplier:  Chemglass Life Sciences CG320050
Encompass_Preferred
1/16" Dia X 12" Long PTFE Coated Thermocouple Probe Type "J". For use with Temperature Controllers CG-15001, CG-3200, CG-3202, CG-3404, CG-3206, CG-3209, and CG-3210. Thermocouples have a standard male plug (OST) and are all PFA COATED.Maximum Probe Temperature Based on Coating- FEP is 200°C and the Black PFA is 250°C
Catalog No. CG320050