Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Tubing, Stainless Steel, 1/16 in. OD, 0.040 in. ID

Supplier:  Waters Corp WAT026805

Encompass_Preferred

This tubing is made of stainless steel and features precise dimensions for scientific and laboratory applications. It has an outer diameter of 1/16 in. and an inner diameter of 0.040 in., supplied in a 10 ft. length.

  • Made of stainless steel
  • Outer diameter of 1/16 in.
  • Inner diameter of 0.040 in.
  • Supplied in a 10 ft. length

Catalog No. 50-785-256


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.