Learn More
Trajan Scientific 1/16IN TO 0.4MM VESPEL FERRULE
Supplier:  Trajan Scientific 212104/DUP
Encompass_Preferred
100% Vespel (registered) polyimide. Temp limit 350 deg. C. 0.4 mm ID fits 0.25 mm ID capillary fused silica tubing. For 1/16 in. Swagelok (registered) / Parker (registered) type fittings. Design requires no back ferrule. Packaged: 3 in. x 3 in. x 1/4 in. Plastic Dial Pack; Weight: 8.5 g
Catalog No. NC9595239