Learn More
Medchemexpress LLC 1,18-Octadecanediol | 3155-43-9 | 286.49 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYN0977
Encompass_Preferred
1,18-Octadecanediol (Octadecane-1,18-diol) is a copolymer of long-chain diol monomers that has properties similar to polyethylene while being easily chemically recyclable and biodegradable.
  • Properties similar to polyethylene.
  • Easily chemically recyclable.
  • Biodegradable.
Catalog No. 50-003-89332