Learn More
Medchemexpress LLC 1-(2-(1-benzyl-2,5-dimethyl-1H-pyrrol-3-yl)-2-oxoethyl)-6-oxo-1,6-dihydropyridine-3-carbonitrile | 1449747-00-5 | MFCD28100815 | 99.5% | 345.39 g/mol | C21H19N3O2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY100953100MG
Encompass_Preferred
CYM-5520 is a selective allosteric agonist of the sphingosine 1-phosphate receptor 2 (S1PR2) supplied for research use. It is a white to off-white solid with reported EC50 ≈ 480 nM and high purity, commonly used in receptor pharmacology and preclinical studies.
  • Selective allosteric S1PR2 agonist with reported EC50 ≈ 480 nM.
  • High purity (99.48%) for reliable experimental results.
  • Supplied as a white to off-white solid suitable for laboratory handling and storage.
  • Available in multiple pack sizes including 100 MG to support varied study scales.
  • Documentation available: data sheet, certificate of analysis, and safety data sheet.
  • Appropriate for in vitro and preclinical receptor signaling studies.
Catalog No. 50-002-12604