Learn More
Medchemexpress LLC 1,2,3,4-Tetramethylbenzene | 488-23-3 | 134.22 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0064161G
Encompass_Preferred
1,2,3,4-Tetramethylbenzene is an aromatic hydrocarbon. It can serve as a molecular marker indicating the input of photosynthetic sulfur bacteria (especially green sulfur bacteria) in ancient sedimentary environments, reflecting the static marine environment of the source rock deposition. It can also be used as a pharmaceutical intermediate.
  • Aromatic hydrocarbon
  • Serves as a molecular marker for photosynthetic sulfur bacteria in ancient sedimentary environments
  • Reflects the static marine environment of source rock deposition
  • Can be used as a pharmaceutical intermediate
  • Exhibits unique regioselectivity in aqueous-phase reaction with F-TEDA-BF4 (Selectfluor)
  • Water molecules attack un-substituted sites on its benzene ring to form a phenol intermediate
  • Does not undergo ipso attack and rearrangement like other highly methylated benzenes
Catalog No. 50-004-47317