Learn More
Medchemexpress LLC 1,2,3,4-Tetramethylbenzene | 488-23-3 | 134.22 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW006416
Encompass_Preferred
1,2,3,4-Tetramethylbenzene is an aromatic hydrocarbon used as a molecular marker for photosynthetic sulfur bacteria in ancient sedimentary environments, indicating static marine conditions. It also functions as a pharmaceutical intermediate. In vitro, it exhibits unique regioselectivity with F-TEDA-BF4 (Selectfluor), where water molecules attack unsubstituted sites on its benzene ring, forming a phenol intermediate.
  • Aromatic hydrocarbon
  • Functions as a molecular marker for photosynthetic sulfur bacteria
  • Can be used as a pharmaceutical intermediate
  • Shows unique regioselectivity in aqueous-phase reactions with F-TEDA-BF4
  • Forms a phenol intermediate from water attacking unsubstituted benzene ring sites
Catalog No. 50-004-48937