Learn More
Cayman Chemical 1 2 3 7 8-Pentachlorodibenzof
SDP
Supplier:  Cayman Chemical 386255 MG
Encompass_Preferred
A a dioxin-like polychlorinated dibenzofuran; induces expression of the genes encoding AHH and EROD in H-4-II-E rat hepatoma cells (EC50s = 2.54 and 3.06 nM, respectively); decreases maternal weight and increases the percentage of mouse fetuses with cleft palates or hydronephrosis in pregnant dams at 100, 150, and 200 UG/kg
Catalog No. 50-291-9016