Learn More
Medchemexpress LLC 1-(2,3-di(thiophen-2-yl)quinoxalin-6-yl)-3-(2-methoxyethyl)urea | 508186-14-9 | MFCD03232237 | 99.7% | 410.51 | C20H18N4O2S2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10403250MG
Encompass_Preferred
Ac-CoA Synthase Inhibitor1 is a research-grade small molecule that reversibly inhibits acetate-dependent acetyl-CoA synthetase 2 (ACSS2). It shows potent activity with an IC50 of 0.6 μM and has reported antiviral activity against respiratory syncytial virus. The compound is provided for biochemical and cellular research in solid form and as a DMSO solution for experimental use.
  • Selective inhibitor of acetyl-CoA synthetase 2 (ACSS2).
  • Potent activity with IC50 0.6 μM.
  • Molecular formula C20H18N4O2S2, molecular weight 410.51.
  • High purity (99.7%).
  • Available as solid and as 10 mM solution in DMSO.
  • Suitable for biochemical and cellular assays.
Catalog No. 50-002-42555