Learn More
Cayman Chemical 1 2 3-TrImyrIstoyl Glycerol 1g
SDP
Supplier:  Cayman Chemical 233371 G
Encompass_Preferred
A triacylglycerol of myristic acid; has been used to prepare lipophilic and cationic lipophilic micro- and nanoparticles carriers loaded with proteins, peptides, and oligonucleotides; has been used as a micro- and nanoparticle carrier of small molecules alone or in combination with tyloxapol; has been used as a substrate to determine fatty acid selectivity of lipases
Catalog No. 50-296-3995