Learn More
Cayman Chemical 1 2 3-TrIoleoyl Glycerol 50g
SDP
Supplier:  Cayman Chemical 2687150 G
Encompass_Preferred
A triacylglycerol; inhibits oxidized LDL-induced decreases in cell viability and SOD and GPX activities and increases in apoptosis in endothelial cells at 10 μM; has been found in sunflower, corn, and extra virgin olive oils; has been used as a standard for the quantification of triacylglycerols in extra virgin olive oil by HTGC-(IT)MS; is a major component of Lorenzo's oil
Catalog No. 50-293-4904