Learn More
Cayman Chemical 1 2 3TrI11 ZEIcosenoyl Gl 10mg
SDP
Supplier:  Cayman Chemical 2692610 MG
Encompass_Preferred
A triacylglycerol; has been used as an internal standard for the quantification of triacylglycerols in seed and olive oils
Catalog No. 50-293-7637