Learn More
Cayman Chemical 1 2 3TrI13 EDocosenoyl Gl
SDP
Supplier:  Cayman Chemical 26991250 MG
Encompass_Preferred
A triacylglycerol; dietary administration at 15% of a calcium-deficient diet transiently increases heart triglyceride levels, which return to baseline after 28 days
Catalog No. 50-293-7121