Learn More
Cayman Chemical 1 2 3TriaLinolenoyl Glycerol
SDP
Supplier:  Cayman Chemical 10009758100 MG
Encompass_Preferred
A polyunsaturated triacylglycerol with α-linolenoyl (18:3) side-chains attached at the C-1, C-2, and C-3 positions that is useful in lipid research
Catalog No. 50-289-0663