Learn More
Cayman Chemical 1 2 3TrIheptnoyl GlycRol 500mg
SDP
Supplier:  Cayman Chemical 28174500 MG
Encompass_Preferred
A triacylglycerol; increases levels of propionyl-, pentanoyl-, and heptanoyl carnitine in mouse blood when administered as 35% of calories in the diet; delays the development of corneal kindling-induced seizures in mice and increases the PTZ tonic extension threshold during the chronically epileptic stage in a mouse model of pilocarpine-induced status epilepticus at 35% of calories in the diet
Catalog No. 50-294-1123