Learn More
Cayman Chemical 1 2 3TrIundecnoyl GlycRol 50mg
SDP
Supplier:  Cayman Chemical 2700850 MG
Encompass_Preferred
A triacylglycerol; dietary administration prevents fasting-induced decreases in plasma glucose, alanine, and insulin levels, as well as the ratio of insulin to glucagon, in rats
Catalog No. 50-293-7119