Learn More
Medchemexpress LLC 1,2,4,5-tetrachloro-3-nitrobenzene | 117-18-0 | >98.0% | 500 MG

Supplier: Medchemexpress LLC HY121935500MG
Tecnazene (1,2,4,5-tetrachloro-3-nitrobenzene) is an organochlorine fungicide used as a sprout inhibitor for stored potatoes and as an intermediate in specialty chemical synthesis. It is supplied for laboratory use in solid powder form and as DMSO solutions for research applications.
- High purity >98.0% (GC).
- Molecular weight 260.89 g·mol⁻¹.
- Chemical formula C6HCl4NO2.
- Available as solids (100 mg, 250 mg, 500 mg, 1 g) and as 10 mM solution in DMSO.
- Suitable for use as a sprout inhibitor and as a building block in specialty chemical synthesis.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.