Learn More
Medchemexpress LLC 1-[2-(4-phenylphenyl)ethyl]-4-[3-(trifluoromethyl)phenyl]-3,6-dihydro-2H-pyridine | 188396-77-2 | MFCD30747895 | 96.3% | 407.47 g·mol⁻¹ | C26H24F3N | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10158025MG
Encompass_Preferred
Paliroden is an orally bioactive, non-peptidic neurotrophic small molecule that activates synthesis of endogenous neurotrophic factors. It has been investigated for potential use in neurodegenerative disorders such as Alzheimer's disease and Parkinson's disease.
  • Orally bioactive small molecule suitable for in vivo studies.
  • Activates synthesis of endogenous neurotrophic factors.
  • Non-peptidic structure improves oral stability.
  • High supplied purity (about 96.3%).
  • Available as a solid, off-white to light yellow powder.
  • Provided in multiple package sizes, including 25 MG.
  • Molecular weight 407.47 g·mol⁻¹; formula C26H24F3N.
Catalog No. 50-002-49112