Learn More
Medchemexpress LLC 1,2,4-Triazolo[4,3-a]pyridin-3(2H)-one | 6969-71-7 | MFCD00022632 | 99.99% | 135.12 g/mol | C6H5N3O | 10 G

Supplier: Medchemexpress LLC HYW05358710G
Triazolopyridinone (3-hydroxytriazolo[4,3-a]pyridine) is a research-grade chemical intermediate used in organic synthesis, notably as a precursor in the synthesis of certain antidepressant derivatives. It is supplied as a high-purity solid for laboratory research use.
- Research-grade chemical intermediate suitable for organic synthesis.
- High purity, reported as 99.99% (HPLC).
- White to off-white solid appearance.
- Chemical formula C6H5N3O, molecular weight 135.12 g/mol.
- Store at room temperature; solutions may be stored at -20°C to -80°C with longer stability at lower temperatures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.