Learn More
Medchemexpress LLC 1,2,4-trideoxy-4-(methylamino)-1-(4-pentylphenyl)-D-erythro-pent-1-enitol hydrochloride | 1072443-89-0 | >=98.0% | 313.86 g/mol | C17H27NO2.HCl | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11901610MG
Encompass_Preferred
SK1-I is a sphingosine analog and a selective, competitive inhibitor of sphingosine kinase 1 (SPHK1) with a reported Ki of approximately 10 μM. It is supplied for research use and is used in biochemical and cell-based studies to modulate sphingolipid signaling, enhance autophagy, and evaluate antitumor activity.
  • Selective SPHK1 inhibitor (Ki ≈ 10 μM).
  • Sphingosine analog used in biochemical and cellular assays.
  • Reported to enhance autophagy and show antitumor effects in preclinical studies.
  • Supplied in small milligram quantities for research use only.
  • Common applications include pathway modulation, proliferation, and apoptosis studies.
Catalog No. 50-003-08201