Learn More
Medchemexpress LLC 1,2,4-trideoxy-4-(methylamino)-1-(4-pentylphenyl)-D-erythro-pent-1-enitol hydrochloride | 1072443-89-0 | >98.0% | 313.86 g/mol | C17H27NO2·HCl | 25 MG

Supplier: Medchemexpress LLC HY11901625MG
SK1-I is a research compound that selectively inhibits sphingosine kinase 1 (SPHK1). It is supplied as the hydrochloride salt (CAS 1072443-89-0) in a 25 mg quantity for laboratory research use only; not for human or veterinary use.
- Selective SPHK1 inhibitor with reported Ki ~10 μM.
- Provided as a hydrochloride salt for improved stability.
- Purity ≥98% (HPLC), suitable for biochemical studies.
- Common applications include cell signaling and anticancer research.
- Store at recommended low temperature to maintain integrity.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.